2-(Methylthio)-5,6,7,8-tetrahydroquinazoline-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A601462-100MG A601462-1G A601462-250MG
77242-592EA 128.75 USD
77242-592 77242-594 77242-596
2-(Methylthio)-5,6,7,8-tetrahydroquinazoline-6-carboxylic acid 97%
2-(Methylthio)-5,6,7,8-tetrahydroquinazoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂S
MW: 224.27 g/mol
MDL Number: MFCD28140851
CAS Number: 1542029-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR