2-Amino-5-bromo-3-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A601049-1G A601049-250MG
77254-246EA 444.96 USD
77254-246 77254-248
2-Amino-5-bromo-3-fluorobenzaldehyde 98%
2-Amino-5-bromo-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MW: 218.02 g/mol
MDL Number: MFCD24445471
CAS Number: 906811-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR