2-Chloro-5-(methoxymethoxy)pyridine 95%

Supplier: Ambeed
Warning

A601046-100MG A601046-1G A601046-250MG A601046-5G
77242-580EA 45.32 USD
77242-580 77242-582 77242-584 77747-238
2-Chloro-5-(methoxymethoxy)pyridine 95%
2-Chloro-5-(methoxymethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
MDL Number: MFCD20278147
CAS Number: 877133-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR