2-Mercapto-1,3,4-thiadiazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,4-Thiadiazole-2-thiol

A600870-25G A600870-5G
77266-698EA 8.24 USD
77266-698 77266-700
2-Mercapto-1,3,4-thiadiazole 98%
2-Mercapto-1,3,4-thiadiazole

Precaución: for Research Use Only

Formula: C₂H₂N₂S₂
MW: 118.17 g/mol
MDL Number: MFCD00040404
CAS Number: 18686-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR