2-((4-Aminophenyl)sulfonyl)ethyl hydrogen sulfate 97%

Supplier: Ambeed
Danger

A600701-25G A600701-5G
77280-240EA 28 USD
77280-240 77280-242
2-((4-Aminophenyl)sulfonyl)ethyl hydrogen sulfate 97%
2-((4-Aminophenyl)sulfonyl)ethyl hydrogen sulfate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₆S₂
MW: 281.30 g/mol
MDL Number: MFCD00100131
CAS Number: 2494-89-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR