(R)-(-)-1-[(S)-2-Di-tert-butylphosphino)ferrocenyl]ethyldi(4-trifluoromethylphenyl)phosphine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-{(RP)-2-[Bis[4-(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi-tert-butylphosphine, (R)-1-{(SP)-2-[Bis[4-(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi-tert-butylphosphine, (S)-1-{(S)-2-[Bis[4-(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi-tert-butylphosphine

A600464-100MG A600464-1G A600464-250MG A600464-50MG A600464-5G
77254-238EA 35.02 USD
77254-238 77254-240 77254-242 77254-244 77747-237
(R)-(-)-1-[(S)-2-Di-tert-butylphosphino)ferrocenyl]ethyldi(4-trifluoromethylphenyl)phosphine 98%
(R)-(-)-1-[(S)-2-Di-tert-butylphosphino)ferrocenyl]ethyldi(4-trifluoromethylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₃₄H₃₈F₆FeP₂
MW: 678.461 g/mol
MDL Number: MFCD04038736
CAS Number: 849924-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR