2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethan-1-one 98%

Supplier: Ambeed
Warning

A600358-1G A600358-25G A600358-5G
77266-692EA 9 USD
77266-692 77266-694 77266-696
2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethan-1-one 98%
2-Bromo-1-(5-fluoro-2-methoxyphenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD03425169
CAS Number: 343-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR