2,2-Difluoroethyl-p-toluenesulfonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2-Difluoroethyl 4-methylbenzenesulphonate
A599728-100MG
A599728-1G
A599728-250MG
A599728-25G
77254-230EA
14.42
USD
77254-230
77254-232
77254-234
77254-236
2,2-Difluoroethyl-p-toluenesulfonate 97%
2,2-Difluoroethyl-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₉H₁₀F₂O₃S MW: 236.24 g/mol |
MDL Number:
MFCD04038284 CAS Number: 135206-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...