2,4-Dichlorobenzaldehyde 98%

Supplier: Ambeed
Danger

A599282-25G A599282-100G
77254-228EA 14 USD
77254-228 77617-514
2,4-Dichlorobenzaldehyde 98%
2,4-Dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O
MDL Number: MFCD00003305
CAS Number: 874-42-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR