5-(Trifluoromethoxy)salicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzoic acid

A598718-1G A598718-250MG A598718-5G
77242-558EA 53.56 USD
77242-558 77242-560 77242-562
5-(Trifluoromethoxy)salicylic acid 98%
5-(Trifluoromethoxy)salicylic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₄
MW: 222.12 g/mol
MDL Number: MFCD01091003
CAS Number: 129644-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR