3-Bromopropionyl chloride 95%

Supplier: Ambeed
Danger

A598176-5G A598176-25G
77280-228EA 15 USD
77280-228 77551-800
3-Bromopropionyl chloride 95%
3-Bromopropionyl chloride

Precaución: for Research Use Only

Formula: BrH₂CCH₂COOCl
MDL Number: MFCD00000746
CAS Number: 15486-96-1
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR