Remdesivir 99%

Supplier: Ambeed
Warning

Synonyms: 2-Ethylbutyl ((S)-(((2R,3S,4R,5R)-5-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)-L-alaninate

A598153-10MG A598153-1G A598153-250MG A598153-25G A598153-5G
77266-684EA 5.15 USD
77266-684 77266-686 77266-688 77266-690 77569-140
Remdesivir 99%
Remdesivir

Precaución: For research use only.

Formula: C₂₇H₃₅N₆O₈P
MW: 602.58 g/mol
MDL Number: MFCD31657351
CAS Number: 1809249-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR