Methyl 2-amino-4-hydroxypyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A597895-100MG A597895-250MG
77242-554EA 78.28 USD
77242-554 77242-556
Methyl 2-amino-4-hydroxypyrimidine-5-carboxylate 97%
Methyl 2-amino-4-hydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₃
MW: 169.13 g/mol
MDL Number: MFCD09880930
CAS Number: 1023815-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR