(S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)propanoic acid 97%

Supplier: Ambeed
Warning

A597473-10G A597473-250MG A597473-25G A597473-5G A597473-1G
77242-546EA 225.57 USD
77242-546 77242-548 77242-550 77242-552 77594-816
(S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)propanoic acid 97%
(S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)propanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₅
MW: 410.46 g/mol
MDL Number: MFCD28387722
CAS Number: 150114-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR