UNC0224 98%

Supplier: Ambeed
Warning

Synonyms: 7-(3-(Dimethylamino)propoxy)-6-methoxy-2-(4-methyl-1,4-diazepan-1-yl)-N-(1-methylpiperidin-4-yl)quinazolin-4-amine

A596599-10MG A596599-1MG A596599-25MG A596599-50MG A596599-5MG
77242-534EA 116 USD
77242-534 77242-536 77242-538 77242-540 77242-542
UNC0224 98%
UNC0224

Precaución: for Research Use Only

Formula: C₂₆H₄₃N₇O₂
MW: 485.67 g/mol
MDL Number: MFCD18251564
CAS Number: 1197196-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR