2,4-Mesitylenedisulfonyl dichloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,4,6-Trimethyl-1,3-benzenedisulphonyl Dichloride, 2,4,6-Trimethylbenzene-1,3-disulphonyl dichloride

A596579-1G A596579-25G A596579-5G
77254-218EA 26.78 USD
77254-218 77254-220 77254-222
2,4-Mesitylenedisulfonyl dichloride 98%
2,4-Mesitylenedisulfonyl dichloride

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O₄S₂
MW: 317.20 g/mol
MDL Number: MFCD00009796
CAS Number: 68985-08-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR