4-(Trimethylsilylethynyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-((Trimethylsilyl)ethynyl)aniline, 4-[(Trimethylsilyl)ethynyl]aniline

A596527-1G A596527-250MG A596527-5G
77292-776EA 44.29 USD
77292-776 77292-778 77292-780
4-(Trimethylsilylethynyl)aniline 98%
4-(Trimethylsilylethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₅NSi
MW: 189.33 g/mol
MDL Number: MFCD03695479
CAS Number: 75867-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR