4,4'-(1H-1,2,4-Triazole-3,5-diyl)dipyridine 97%

Supplier: Ambeed
Warning

A596060-100MG A596060-1G A596060-250MG A596060-5G
77254-210EA 7 USD
77254-210 77254-212 77254-214 77254-216
4,4'-(1H-1,2,4-Triazole-3,5-diyl)dipyridine 97%
4,4'-(1H-1,2,4-Triazole-3,5-diyl)dipyridine

Precaución: for Research Use Only

Formula: C₁₂H₉N₅
MW: 223.23 g/mol
MDL Number: MFCD06297778
CAS Number: 4329-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR