4-Phenyl-1-butyne 98%

Supplier: Ambeed
Warning

A595890-1G A595890-25G
77292-766EA 46.35 USD
77292-766 77292-768
4-Phenyl-1-butyne 98%
4-Phenyl-1-butyne

Precaución: for Research Use Only

Formula: C₁₀H₁₀
MW: 130.19 g/mol
MDL Number: MFCD00041667
CAS Number: 16520-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR