4,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan ≥98%, ee 99%

Supplier: Ambeed
Warning

A595796-100MG A595796-1G A595796-250MG A595796-50MG
77266-676EA 87.55 USD
77266-676 77266-678 77266-680 77266-682
4,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan ≥98%, ee 99%
4,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₃₀H₂₂N₂O₃
MW: 458.50 g/mol
MDL Number: MFCD30471362
CAS Number: 195433-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR