1-Bromo-9-phenyl-9H-carbazole 97%

Supplier: Ambeed
Warning

A595296-1G A595296-250MG
77242-524EA 63.86 USD
77242-524 77242-526
1-Bromo-9-phenyl-9H-carbazole 97%
1-Bromo-9-phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂BrN
MW: 322.21 g/mol
MDL Number: MFCD28975084
CAS Number: 1333002-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR