Methyl 2-methoxy-5-methylbenzoate 97%

Supplier: Ambeed
Warning

A595048-1G A595048-5G
77242-520EA 18 USD
77242-520 77242-522
Methyl 2-methoxy-5-methylbenzoate 97%
Methyl 2-methoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD06411343
CAS Number: 63113-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR