2-Fluoro-6-iodobenzyl bromide 97%

Supplier: Ambeed
Warning

A594591-1G A594591-5G
77266-670EA 47 USD
77266-670 77266-672
2-Fluoro-6-iodobenzyl bromide 97%
2-Fluoro-6-iodobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MDL Number: MFCD18396957
CAS Number: 1231747-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR