4-Chloro-6,7-dimethoxy-2-methylquinoline 95%

Supplier: Ambeed
Warning

A594498-100MG A594498-250MG
77266-666EA 65 USD
77266-666 77266-668
4-Chloro-6,7-dimethoxy-2-methylquinoline 95%
4-Chloro-6,7-dimethoxy-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClNO₂
MDL Number: MFCD11868302
CAS Number: 100122-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR