1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1S,2S)-2-(dimethylamino)cyclohexyl)urea 98%

Supplier: Ambeed
Warning

A594470-250MG A594470-1G A594470-100MG
77254-208EA 167 USD
77254-208 77617-502 77747-230
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1S,2S)-2-(dimethylamino)cyclohexyl)urea 98%
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1S,2S)-2-(dimethylamino)cyclohexyl)urea

Precaución: for Research Use Only

Formula: C₁₇H₂₁F₆N₃O
MDL Number: MFCD23380084
CAS Number: 1221442-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR