4-Fluoro-2-methylbenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Fluoro-o-toluoyl chloride, 2-(Chlorocarbonyl)-5-fluorotoluene

A594103-25G A594103-5G
77292-756EA 53 USD
77292-756 77292-758
4-Fluoro-2-methylbenzoyl chloride 98%
4-Fluoro-2-methylbenzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MDL Number: MFCD04115881
CAS Number: 21900-43-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR