4,6-Dichlorobenzofuran 95%

Supplier: Ambeed
Warning

A593959-100MG A593959-1G A593959-250MG
77292-750EA 26 USD
77292-750 77292-752 77292-754
4,6-Dichlorobenzofuran 95%
4,6-Dichlorobenzofuran

Precaución: for Research Use Only

Formula: C₈H₄Cl₂O
MDL Number: MFCD00052494
CAS Number: 175203-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR