p-Propylanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-4-propylbenzene, 4-Propylanisole

A593578-100G A593578-25G A593578-500G A593578-5G
77292-742EA 53.56 USD
77292-742 77292-744 77292-746 77292-748
p-Propylanisole 97%
p-Propylanisole

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD00027121
CAS Number: 104-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR