2-Fluoro-6-nitrophenol 98%

Supplier: Ambeed
Danger

A592854-100G A592854-10G A592854-1G A592854-25G A592854-5G
77292-732EA 113 USD
77292-732 77292-734 77292-736 77292-738 77292-740
2-Fluoro-6-nitrophenol 98%
2-Fluoro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄FNO₃
MDL Number: MFCD00042446
CAS Number: 1526-17-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR