Methyl-3-(bromomethyl)benzoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 3-(bromomethyl)benzoate

A592293-100G A592293-10G A592293-1G A592293-500G A592293-5G A592293-25G
77266-652EA 155 USD
77266-652 77266-654 77266-656 77266-658 77266-660 77551-786
Methyl-3-(bromomethyl)benzoate 98%
Methyl 3-(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00051437
CAS Number: 1129-28-8
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR