(3-Aminopropyl)dimethylethoxysilane 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Ethoxydimethylsilyl)propan-1-amine

A592249-1G A592249-5G A592249-25G
77280-218EA 24.72 USD
77280-218 77280-220 77572-894
(3-Aminopropyl)dimethylethoxysilane 95%
(3-Aminopropyl)dimethylethoxysilane

Precaución: for Research Use Only

Formula: C₇H₁₉NOSi
MW: 161.3174 g/mol
MDL Number: MFCD00053947
CAS Number: 18306-79-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR