WS-12 97%

Supplier: Ambeed
Warning

Synonyms: AVX-012, (1R,2S,5R)-2-Isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexane-1-carboxamide

A591847-10MG A591847-1MG A591847-5MG
77266-642EA 139.05 USD
77266-642 77266-644 77266-646
WS-12 97%
WS-12

Precaución: for Research Use Only

Formula: C₁₈H₂₇NO₂
MW: 289.4 g/mol
MDL Number: MFCD11100191
CAS Number: 68489-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR