4-Amino-2-methylphenylboronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-2-methylphenylboronic acid, pinacol ester, 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
A591741-100MG
A591741-250MG
A591741-5G
A591741-1G
77292-726EA
35.02
USD
77292-726
77292-728
77292-730
77569-126
4-Amino-2-methylphenylboronic acid pinacol ester 96%
4-Amino-2-methylphenylboronic acid pinacol ester
Precaución: For research use only.
Formula:
C₁₃H₂₀BNO₂ MW: 233.12 g/mol |
MDL Number:
MFCD08458198 CAS Number: 631911-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...