Benzo[c][1,2]oxaborole-1,5(3H)-diol 97%

Supplier: Ambeed
Warning

A591695-100MG A591695-1G A591695-250MG A591695-5G
77266-634EA 30 USD
77266-634 77266-636 77266-638 77266-640
Benzo[c][1,2]oxaborole-1,5(3H)-diol 97%
Benzo[c][1,2]oxaborole-1,5(3H)-diol

Precaución: for Research Use Only

Formula: C₇H₇BO₃
MDL Number: MFCD22393581
CAS Number: 1187190-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR