1-Methyl-3-propylimidazolium bromide 98%

Supplier: Ambeed
Warning

Synonyms: 1-propenyl-3-methylimidazolium bromide

A591402-25G A591402-5G
77266-630EA 27.81 USD
77266-630 77266-632
1-Methyl-3-propylimidazolium bromide 98%
1-Methyl-3-propylimidazolium bromide

Precaución: for Research Use Only

Formula: C₇H₁₃BrN₂
MW: 205.10 g/mol
MDL Number: MFCD17014727
CAS Number: 85100-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR