2-Dimethylaminoethyl benzoate 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Dimethylamino)ethyl benzoate, Benzoic Acid 2-Dimethylaminoethyl Ester

A591195-10G A591195-25G A591195-5G
77254-198EA 17.51 USD
77254-198 77254-200 77254-202
2-Dimethylaminoethyl benzoate 97%
2-Dimethylaminoethyl benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
MDL Number: MFCD00051067
CAS Number: 2208-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR