1,2-Butylene carbonate 98%

Supplier: Ambeed
Warning

A591099-100G A591099-25G A591099-500G A591099-5G
77292-714EA 184 USD
77292-714 77292-716 77292-718 77292-720
1,2-Butylene carbonate 98%
1,2-Butylene carbonate

Precaución: for Research Use Only

Formula: C₅H₈O₃
MDL Number: MFCD00143317
CAS Number: 4437-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR