Adavivint 97%

Supplier: Ambeed
Warning

Synonyms: N-(5-{3-[7-(3-fluorophenyl)-1H-imidazo[4,5-c]pyridin-2-yl]-1H-indazol-5-yl}pyridin-3-yl)-3-methylbutanamide, SM04690, N-(5-(3-(7-(3-Fluorophenyl)-3H-imidazo[4,5-c]pyridin-2-yl)-1H-indazol-5-yl)pyridin-3-yl)-3-methylbutanamide

A591020-10MG A591020-1MG A591020-25MG A591020-5MG
77254-190EA 121 USD
77254-190 77254-192 77254-194 77254-196
Adavivint 97%
Adavivint

Precaución: for Research Use Only

Formula: C₂₉H₂₄FN₇O
MDL Number: MFCD30489411
CAS Number: 1467093-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR