4,4',4'',4'''-Silanetetrayltetrabenzaldehyde 97%

Supplier: Ambeed
Warning

A590515-100MG A590515-250MG
77266-626EA 23 USD
77266-626 77266-628
4,4',4'',4'''-Silanetetrayltetrabenzaldehyde 97%
4,4',4'',4'''-Silanetetrayltetrabenzaldehyde

Precaución: for Research Use Only

Formula: C₂₈H₂₀O₄Si
MW: 448.54 g/mol
MDL Number: MFCD31631360
CAS Number: 1055999-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR