1-(2-Ethoxy-2-oxoethyl)pyridinium bromide 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Ethoxy-2-oxoethyl)pyridin-1-ium bromide

A590510-100MG A590510-250MG A590510-1G
77254-186EA 20 USD
77254-186 77254-188 77594-798
1-(2-Ethoxy-2-oxoethyl)pyridinium bromide 98%
1-(2-Ethoxy-2-oxoethyl)pyridinium bromide

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO₂
MDL Number: MFCD00463694
CAS Number: 17282-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR