1,4-Diacetoxy-2-butyne 98%

Supplier: Ambeed
Warning

Synonyms: But-2-yne-1,4-diyl diacetate, 2-Butyne-1,4-diol Diacetate, Bis(acetoxymethyl)acetylene

A590243-1G A590243-25G A590243-5G
77266-620EA 11.33 USD
77266-620 77266-622 77266-624
1,4-Diacetoxy-2-butyne 98%
1,4-Diacetoxy-2-butyne

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MW: 170.16 g/mol
MDL Number: MFCD00014982
CAS Number: 1573-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR