1-Ethyl-2-methyl-1H-imidazole 98%

Supplier: Ambeed
Warning

A590197-100G A590197-25G A590197-5G
77266-614EA 107.12 USD
77266-614 77266-616 77266-618
1-Ethyl-2-methyl-1H-imidazole 98%
1-Ethyl-2-methyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₆H₁₀N₂
MW: 110.157 g/mol
MDL Number: MFCD18840475
CAS Number: 21202-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR