tert-Butyl 2-((tert-butoxycarbonyl)amino)-2-cyanoacetate 97%

Supplier: Ambeed
Warning

A590156-100MG A590156-250MG
77242-506EA 14.42 USD
77242-506 77242-508
tert-Butyl 2-((tert-butoxycarbonyl)amino)-2-cyanoacetate 97%
tert-Butyl 2-((tert-butoxycarbonyl)amino)-2-cyanoacetate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₄
MW: 256.3 g/mol
MDL Number: MFCD24466150
CAS Number: 130892-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR