4-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-ethoxycarbonylphenylboronic acid

A590019-100MG A590019-1G A590019-250MG A590019-5G
77254-178EA 36.05 USD
77254-178 77254-180 77254-182 77254-184
4-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%
4-Chloro-(2-ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BClO₄
MW: 228.4400 g/mol
MDL Number: MFCD06659823
CAS Number: 850568-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR