(4-Methoxy-3,5-dimethylpyridin-2-yl)methanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A589550-100MG A589550-1G A589550-250MG
77242-500EA 28 USD
77242-500 77242-502 77242-504
(4-Methoxy-3,5-dimethylpyridin-2-yl)methanamine dihydrochloride 97%
(4-Methoxy-3,5-dimethylpyridin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆Cl₂N₂O
MDL Number: MFCD12026975
CAS Number: 1158207-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR