2-(3-Thienyl)pyridine 98+%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Pyridyl)thiophene, 2-(Thiophen-3-yl)pyridine

A589520-100MG A589520-1G A589520-250MG A589520-5G
77242-492EA 15.45 USD
77242-492 77242-494 77242-496 77242-498
2-(3-Thienyl)pyridine 98+%
2-(3-Thienyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₇NS
MW: 161.22 g/mol
MDL Number: MFCD00006214
CAS Number: 21298-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR