3-Bromo-2-isopropylpyridine 97%

Supplier: Ambeed
Warning

A589441-100MG A589441-250MG A589441-50MG
77266-604EA 167 USD
77266-604 77266-606 77266-608
3-Bromo-2-isopropylpyridine 97%
3-Bromo-2-isopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MDL Number: MFCD23143906
CAS Number: 1417518-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR