1-Methyl-2-phenyl-1H-indole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A589435-1G A589435-25G A589435-5G
77280-208EA 12.36 USD
77280-208 77280-210 77280-212
1-Methyl-2-phenyl-1H-indole-3-carbaldehyde 98%
1-Methyl-2-phenyl-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO
MW: 235.28 g/mol
MDL Number: MFCD00186065
CAS Number: 1757-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR