3-Furaldehyde 97%

Supplier: Ambeed
Danger

Synonyms: Furan-3-carbaldehyde

A589248-10G A589248-1G A589248-25G A589248-5G
77280-198EA 94.76 USD
77280-198 77280-200 77280-202 77280-204
3-Furaldehyde 97%
3-Furaldehyde

Precaución: for Research Use Only

Formula: C₅H₄O₂
MW: 96.09 g/mol
MDL Number: MFCD00010424
CAS Number: 498-60-2
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR