2-Chloro-6-methylbenzonitrile 97%
Supplier: Ambeed
|
Danger
|
A589112-5G
A589112-25G
77266-594EA
104.03
USD
77266-594
77749-211
2-Chloro-6-methylbenzonitrile 97%
2-Chloro-6-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆ClN MW: 151.60 g/mol |
MDL Number:
MFCD00001782 CAS Number: 6575-09-3 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...